2-Ethylhexane-1,3-diol (mixture of isomers)

Supplier: TCI America

E0119-25ML E0119-500ML
TCE0119-025MLEA 37.15 CAD
TCE0119-025ML TCE0119-500ML
2-Ethylhexane-1,3-diol (mixture of isomers)
2-Ethylhexane-1,3-diol (mixture of isomers)

CAS Number: 94-96-2
MDL Number: MFCD00004578
Molecular Formula: C8H18O2
Molecular Weight: 146.23
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 244
Melting point (°C): -40
Flash Point (°C): 130
Specific Gravity (20/20): 0.94

Formula: C₈H₁₈O₂
Boiling Pt: 244 °C
Melting Pt: –40 °C
Density: 0.94 (20 °C)
Flash Pt: 130 °C
MDL Number: MFCD00004578
CAS Number: 94-96-2
EINECS: 202-377-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=462e38cde787d330d843f044f099463121b31a05&vid=462e38cde787d330d843f044f099463121b31a05&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR