Ethylene di(acetate) ≥97.0%

Supplier: TCI America

Synonyms: Ethane-1,2-diyl diacetate, Ethylene glycol diacetate, Ethylene Glycol Diacetate, 1,2-Diacetoxyethane

E0109-25ML E0109-500ML
TCE0109-25MLEA 33.18 CAD
TCE0109-25ML TCE0109-500ML
Ethylene di(acetate) ≥97.0%
Ethylene di(acetate)

CAS Number: 111-55-7
MDL Number: MFCD00008718
Molecular Formula: C6H10O4
Molecular Weight: 146.14
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 190
Flash Point (°C): 96
Freezing point (°C): -31
Specific Gravity (20/20): 1.11


Other Product Names / Synonyms: 1,2-Diacetoxyethane, 1,2-Ethanediol Diacetate, Ethylene Diacetate, Glycol Diacetate

Formula: C₆H₁₀O₄
MW: 146.14 g/mol
Boiling Pt: 190 °C
Melting Pt: –31 °C
Density: 1.11 (20 °C)
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00008718
CAS Number: 111-55-7
EINECS: 203-881-1
Merck Index: 13,03833
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2db99fca7896036a0df989f24f27f2e829565eb6&vid=2db99fca7896036a0df989f24f27f2e829565eb6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR