Ethylene glycol ≥99.5%

Supplier: TCI America
Warning

E0105-500G E0105-25G
TCE0105-500GEA 58.89 CAD
TCE0105-500G TCE0105-025G
Ethylene glycol ≥99.5%
Ethylene glycol

CAS Number: 107-21-1
MDL Number: MFCD00002885
Molecular Formula: C2H6O2
Molecular Weight: 62.07
Purity/Analysis Method: >99.5% (GC)
Form: Clear Liquid
Boiling point (°C): 196
Melting point (°C): -13
Flash Point (°C): 127
Specific Gravity (20/20): 1.12


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,2-Dihydroxyethane

Formula: HOCH₂CH₂OH
Boiling Pt: 196 °C
Melting Pt: –13 °C
Density: 1.12 (20 °C)
Flash Pt: 127 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002885
CAS Number: 107-21-1
EINECS: 203-473-3
REACH: 01-2119456816-28
Merck Index: 13,03832
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f4c9f22821b86612a45ea543a96bcc9bce991f02&vid=ac6801d3f1a132d01ff47f2546f78a07757fc8c4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.5% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR