Ethylene glycol dimethacrylate (ethylene dimethacrylate) ≥97.0% stabilised

Supplier: TCI America

Synonyms: Ethylene dimethacrylate, Ethane-1,2-diyl bis(2-methylacrylate)

E0102-500ML E0102-25ML
TCE0102-500MLEA 176.39 CAD
TCE0102-500ML TCE0102-025ML
Ethylene glycol dimethacrylate (ethylene dimethacrylate) ≥97.0% stabilised
Ethylene glycol dimethacrylate (Ethylene dimethacrylate)

(stabilized with HQ)
CAS Number: 97-90-5
MDL Number: MFCD00008590
Molecular Formula: C10H14O4
Molecular Weight: 198.22
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 235
Melting point (°C): -40
Flash Point (°C): 118
Specific Gravity (20/20): 1.05


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Ethylene Dimethacrylate, Glycol Dimethacrylate

Formula: C₁₀H₁₄O₄
MW: 198.21 g/mol
Boiling Pt: 235 °C
Melting Pt: –40 °C
Density: 1.05 (20 °C)
Flash Pt: 118 °C
MDL Number: MFCD00008590
CAS Number: 97-90-5
EINECS: 202-617-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b6f167cdd3b978b71f1eccec1732b8b4506205db&vid=b6f167cdd3b978b71f1eccec1732b8b4506205db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR