EDTA ferric monosodium salt ≥90.0%

Supplier: TCI America
Warning

Synonyms: Ethylenedinitrilotetra-acetic acid iron (III) sodium salt, Edetic acid iron (III) monosodium salt, Edetic acid iron (III) sodium salt, Ethylenediaminetetra-acetic acid iron (III) sodium salt, Iron (III) sodium ethylenediaminetetraacetate, Ethylenedinitrilotetra-acetic acid iron (III) monosodium salt, EDTA iron sodium, Sodium ferric ethylenediaminetetraacetate, Iron (III) monosodium ethylenediaminetetraacetate, Sodium feredetate, Ethylenediaminetetra-acetic acid iron (III) monosodium salt

E0092-25G E0092-500G
TCE0092-25GEA 56.44 CAD
TCE0092-25G TCE0092-500G
EDTA ferric monosodium salt ≥90.0%
EDTA ferric monosodium salt

CAS Number: 15708-41-5
MDL Number: MFCD00078215
Molecular Formula: C10H16N2O8
Molecular Weight: 367.05
Purity/Analysis Method: >90.0% (T)
Form: Crystal


Other Product Names / Synonyms: Ethylenediaminetetraacetic Acid Ferric Sodium Salt, Ethylenediaminetetraacetic Acid Iron(III) Sodium Salt, Ferric Sodium Ethylenediaminetetraacetate, Iron(III) Sodium Ethylenediaminetetraacetate

Formula: C₁₀H₁₂FeN₂NaO₈
MW: 367.05 g/mol
Melting Pt: 80
Storage Temperature: Ambient
MDL Number: MFCD00078215
CAS Number: 15708-41-5
EINECS: 239-802-2
Merck Index: 12,04076
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38c1cd68065264e89e88f7d0d33492570c5365f1&vid=38c1cd68065264e89e88f7d0d33492570c5365f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR