EDTA disodium salt dihydrate ≥99.5%

Supplier: TCI America

Synonyms: Ethylenediaminetetraacetic acid, disodium salt, dihydrate, Ethylenediaminetetraacetic acid disodium salt dihydrate, Kestranal® 2S dihydrate, Disodium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenedinitrilotetra-acetic acid disodium salt dihydrate, Ethylenediaminetetraacetic acid disodium dihydrate, Titriplex III, Ethylenediaminetetra-acetic acid disodium salt dihydrate, Kestranal® 2S 2 H2O, Sequestrene Na₂, Edetate disodium dihydrate, Edetic acid disodium salt dihydrate

E0091-500G E0091-25G
TCE0091-500GEA 208.51 CAD
TCE0091-500G TCE0091-025G
EDTA disodium salt dihydrate ≥99.5%
EDTA disodium salt dihydrate

CAS Number: 6381-92-6
MDL Number: MFCD00150037
Molecular Formula: C10H16N2O8
Molecular Weight: 336.21
Purity/Analysis Method: >99.5% (T)
Form: Crystal
Melting point (°C): 252


Product Categories: Analytical Science, Complexones (Chelating Agents)
Other Product Names / Synonyms: Ethylenediaminetetraacetic Acid Disodium Salt, EDTA 2Na, ETA

Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
MW: 372.24 g/mol
Melting Pt: 255
Storage Temperature: Ambient
MDL Number: MFCD00150037
CAS Number: 6381-92-6
EINECS: 205-358-3
Merck Index: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e230b41a3f864c1cbde8d65426bfb85b27ba4c6&vid=4e230b41a3f864c1cbde8d65426bfb85b27ba4c6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.5% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR