2-Ethyl-9,10-anthraquinone ≥98.0%

Supplier: TCI America

Synonyms: 2-Ethylanthracene-9,10-dione, 2-Ethylanthraquinone

E0063-500G E0063-25G
TCE0063-500GEA 255.96 CAD
TCE0063-500G TCE0063-025G
2-Ethyl-9,10-anthraquinone ≥98.0%
2-Ethyl-9,10-anthraquinone

CAS Number: 84-51-5
MDL Number: MFCD00001237
Molecular Formula: C16H12O2
Molecular Weight: 236.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 190
Melting point (°C): 108
Flash Point (°C): 210

Formula: C₁₆H₁₂O₂
MW: 236.27 g/mol
Boiling Pt: 190 °C
Melting Pt: 108 °C
Flash Pt: 210 °C
MDL Number: MFCD00001237
CAS Number: 84-51-5
EINECS: 201-535-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aba3697594347a5d3c81fbc851e5f121982ca1e2&vid=aba3697594347a5d3c81fbc851e5f121982ca1e2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR