3-Ethoxyphenol ≥97.0%

Supplier: TCI America
Warning

E0051-25ML E0051-5ML
TCE0051-025MLEA 401.32 CAD
TCE0051-025ML TCE0051-5ML
3-Ethoxyphenol ≥97.0%
3-Ethoxyphenol

CAS Number: 621-34-1
MDL Number: MFCD00016450
Molecular Formula: C8H10O2
Molecular Weight: 138.17
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 247
Freezing point (°C): 31


Other Product Names / Synonyms: Resorcinol Monoethyl Ether

Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 131
Density: 1.105 g/cm³ (15 °C)
MDL Number: MFCD00016450
CAS Number: 621-34-1
EINECS: 210-681-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f81c678db6ae58a7287c1ec7df72fff41e98479&vid=7f81c678db6ae58a7287c1ec7df72fff41e98479&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR