Ethylvanillin ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

E0050-500G E0050-25G
TCE0050-500GEA 205.4 CAD
TCE0050-500G TCE0050-025G
Ethylvanillin ≥98.0% (by HPLC, titration analysis)
Ethylvanillin

CAS Number: 121-32-4
MDL Number: MFCD00006944
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 78
Flash Point (°C): 100


Other Product Names / Synonyms: Ethyl Vanillin

Formula: C₉H₁₀O₃
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 78 °C
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00006944
CAS Number: 121-32-4
EINECS: 204-464-7
Merck Index: 13,03890
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac3f5ce883597b04dcf92f3f93c73a1f92e6a9b9&vid=983d7d4ac7c3baf2dffd72e5ab6d2380d89513f0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR