Diethylene glycol monoethyl ether ≥99.0%

Supplier: TCI America

E0048-25G E0048-500G
TCE0048-25GEA 41.79 CAD
TCE0048-25G TCE0048-500G
Diethylene glycol monoethyl ether ≥99.0%
Diethylene glycol monoethyl ether

CAS Number: 111-90-0
MDL Number: MFCD00002872
Molecular Formula: C6H14O3
Molecular Weight: 134.18
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 196
Flash Point (°C): 96
Specific Gravity (20/20): 0.99


Other Product Names / Synonyms: Carbitol, 2-(2-Ethoxyethoxy)ethanol

Formula: C₆H₁₄O₃
Boiling Pt: 196 °C
Melting Pt: –80 °C
Density: 0.99 (20 °C)
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00002872
CAS Number: 111-90-0
EINECS: 203-919-7
REACH: 01-2119475105-42
Merck Index: 13,01809
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc829667474b9059ba745465d2456f6ac6f3ad6a&vid=fc829667474b9059ba745465d2456f6ac6f3ad6a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR