4-Ethoxybenzaldehyde ≥98.0%

Supplier: TCI America
Warning

E0042-25G E0042-500G
TCE0042-025GEA 61.62 CAD
TCE0042-025G TCE0042-500G
4-Ethoxybenzaldehyde ≥98.0%
4-Ethoxybenzaldehyde

CAS Number: 10031-82-0
MDL Number: MFCD00003388
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 255
Flash Point (°C): 75
Freezing point (°C): 14
Specific Gravity (20/20): 1.08

Formula: C₉H₁₀O₂
Boiling Pt: 255 °C
Melting Pt: 13…14 °C
Density: 1.08 (20 °C)
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00003388
CAS Number: 10031-82-0
EINECS: 233-093-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18d64556c5e6b4b691451178a55f10ea9408e629&vid=f441f063b0077fdb03054480cd963fa5ed0842a4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR