Arachidyl alcohol ≥95.0%

Supplier: TCI America

E0004-500G E0004-25G
TCE0004-500GEA 448.72 CAD
TCE0004-500G TCE0004-025G
Arachidyl alcohol ≥95.0%
Arachidyl alcohol

CAS Number: 629-96-9
MDL Number: MFCD00002938
Molecular Formula: C20H42O
Molecular Weight: 298.56
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 190
Melting point (°C): 65

Formula: H₃C(CH₂)₁₈CH₂OH
Boiling Pt: 190 °C
Melting Pt: 65 °C
Density: 0.88 g/cm³ (21 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002938
CAS Number: 629-96-9
EINECS: 211-119-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62240dec5a0eb626785cccc7444df1ae4bdc547e&vid=4e676cafecc3d7b85dad639e7c754728f8552d68&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR