EDTA dipotassium salt dihydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D3901-25G D3901-500G
TCD3901-25GEA 44.24 CAD
TCD3901-25G TCD3901-500G
EDTA dipotassium salt dihydrate ≥98.0% (by titrimetric analysis)
EDTA dipotassium salt dihydrate

CAS Number: 25102-12-9
MDL Number: MFCD00150036
Molecular Formula: C10H16N2O8
Molecular Weight: 368.42
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 272


Product Categories: Analytical Science, Complexones (Chelating Agents)
Other Product Names / Synonyms: Ethylenediaminetetraacetic Acid Dipotassium Salt, EDTA 2K

Formula: C₁₀H₁₄K₂N₂O₈·2H₂O
Melting Pt: 272 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150036
CAS Number: 25102-12-9
EINECS: 217-895-0
Merck Index: 13,03546
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75e9a340c7d608cf4a7dc127ff414548db8d4d96&vid=75e9a340c7d608cf4a7dc127ff414548db8d4d96&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR