(+)-Dibenzoyl-D-tartaric acid ≥98.0%

Supplier: TCI America

Synonyms: (2S,3S)-(+)-Di-O-benzoyltartaric acid, (2S,3S)-2,3-Bis(benzoyloxy)succinic acid

D3826-250G D3826-25G
TCD3826-250GEA 145.36 CAD
TCD3826-250G TCD3826-25G
(+)-Dibenzoyl-D-tartaric acid ≥98.0%
(+)-Dibenzoyl-D-tartaric acid

CAS Number: 17026-42-5
MDL Number: MFCD00063222
Molecular Formula: C18H14O8
Molecular Weight: 358.30
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 154
Specific rotation [a]20/D: 117 deg (C=1, EtOH)

Formula: C₁₈H₁₄O₈
MW: 358.31 g/mol
Melting Pt: 154 °C
MDL Number: MFCD00063222
CAS Number: 17026-42-5
EINECS: 241-097-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82c4cf0481c294d5f77230a856ece4ac28e42c2b&vid=82c4cf0481c294d5f77230a856ece4ac28e42c2b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR