1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

D3611-500MG D3611-5G D3611-25G
TCD3611-500MGEA 86.81 CAD
TCD3611-500MG TCD3611-5G TCD3611-25G
1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride ≥98.0% (by HPLC, titration analysis)
1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride

CAS Number: 250285-32-6
MDL Number: MFCD02684545
Molecular Formula: C27H37ClN2
Molecular Weight: 425.06
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 278


Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, Ligands
Other Product Names / Synonyms: IPr.HCl

Formula: C₂₇H₃₇ClN₂
Melting Pt: 278 °C
MDL Number: MFCD02684545
CAS Number: 250285-32-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ece60b8e5bb1008cf1c730c5eb465269031821e7&vid=ece60b8e5bb1008cf1c730c5eb465269031821e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR