Dodecyltrimethoxysilane ≥93.0%

Supplier: TCI America

D3383-25G D3383-250G
TCD3383-25GEA 67.94 CAD
TCD3383-25G TCD3383-250G
Dodecyltrimethoxysilane ≥93.0%
Dodecyltrimethoxysilane

CAS Number: 3069-21-4
MDL Number: MFCD00069148
Molecular Formula: C15H34O3Si
Molecular Weight: 290.52
Purity/Analysis Method: >93.0% (GC)
Form: Clear Liquid
Boiling point (°C): 118
Melting point (°C): -40
Flash Point (°C): 108
Specific Gravity (20/20): 0.90


Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: Lauryltrimethoxysilane

Formula: C₁₅H₃₄O₃Si
Boiling Pt: 118 °C
Melting Pt: –40 °C
Density: 0.90 (20 °C)
Flash Pt: 108 °C
MDL Number: MFCD00069148
CAS Number: 3069-21-4
EINECS: 221-332-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9db404294a3ea869083a658236ca72f38014d9c0&vid=9db404294a3ea869083a658236ca72f38014d9c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR