Diethoxy(diphenyl)silane ≥98.0%
Supplier: TCI America
Synonyms:
Diphenyldiethoxysilane, Diethoxydiphenylsilane
D3067-25G
D3067-100G
TCD3067-25GEA
123.24
CAD
TCD3067-25G
TCD3067-100G
Diethoxy(diphenyl)silane ≥98.0%
Diethoxy(diphenyl)silane
CAS Number: 2553-19-7
MDL Number: MFCD00015126
Molecular Formula: C16H20O2Si
Molecular Weight: 272.42
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 218
Flash Point (°C): 175
Specific Gravity (20/20): 1.03
Formula:
C₁₆H₂₀O₂Si MW: 272.42 g/mol Boiling Pt: 218 °C Density: 1.03 (20 °C) Flash Pt: 175 °C |
MDL Number:
MFCD00015126 CAS Number: 2553-19-7 EINECS: 219-860-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...