2'-Hydroxy-4',6'-dimethoxyacetophenone ≥98.0%
Supplier: TCI America
D2683-1G
D2683-5G
TCD2683-001GEA
77.42
CAD
TCD2683-001G
TCD2683-005G
2'-Hydroxy-4',6'-dimethoxyacetophenone ≥98.0%
2'-Hydroxy-4',6'-dimethoxyacetophenone
CAS Number: 90-24-4
MDL Number: MFCD00017243
Molecular Formula: C10H12O4
Molecular Weight: 196.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 185
Melting point (°C): 81
Other Product Names / Synonyms: 2'-Hydroxy-4',6'-dimethoxyacetophenone
Formula:
C₁₀H₁₂O₄ Boiling Pt: 185 °C Melting Pt: 81 °C |
MDL Number:
MFCD00017243 CAS Number: 90-24-4 EINECS: 201-978-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...