3',5'-Dimethoxy-4'-hydroxyacetophenone ≥98.0%

Supplier: TCI America

D2666-5G D2666-25G
TCD2666-005GEA 279.38 CAD
TCD2666-005G TCD2666-025G
3',5'-Dimethoxy-4'-hydroxyacetophenone ≥98.0%
3',5'-Dimethoxy-4'-hydroxyacetophenone

CAS Number: 2478-38-8
MDL Number: MFCD00008748
Molecular Formula: C10H12O4
Molecular Weight: 196.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 126


Other Product Names / Synonyms: Acetosyringone

Formula: C₁₀H₁₂O₄
Boiling Pt: 335 °C (1013 hPa)
Melting Pt: 126 °C
MDL Number: MFCD00008748
CAS Number: 2478-38-8
EINECS: 219-610-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e4334498dd06afab9c0e8c4ab1408fca83323de&vid=8e4334498dd06afab9c0e8c4ab1408fca83323de&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR