3,5-Di-tert-butylbenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D2663-25G D2663-5G D2663-1G
TCD2663-25GEA 540.52 CAD
TCD2663-25G TCD2663-005G TCD2663-001G
3,5-Di-tert-butylbenzoic acid ≥98.0% (by GC, titration analysis)
3,5-Di-tert-butylbenzoic acid

CAS Number: 16225-26-6
MDL Number: MFCD00082727
Molecular Formula: C15H22O2
Molecular Weight: 234.34
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 173

Formula: C₁₅H₂₂O₂
Melting Pt: 173 °C
MDL Number: MFCD00082727
CAS Number: 16225-26-6
EINECS: 240-350-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b3917eb5ece337215283630595b0dfe8a734b77a&vid=b3917eb5ece337215283630595b0dfe8a734b77a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR