5-(Hydroxymethyl)resorcinol ≥97.0%

Supplier: TCI America

Synonyms: 3,5-Dihydroxybenzyl alcohol

D2656-5G D2656-25G
TCD2656-005GEA 165.9 CAD
TCD2656-005G TCD2656-25G
5-(Hydroxymethyl)resorcinol ≥97.0%
5-(Hydroxymethyl)resorcinol

CAS Number: 29654-55-5
MDL Number: MFCD00016867
Molecular Formula: C7H8O3
Molecular Weight: 140.14
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 186

Formula: C₇H₈O₃
MW: 140.14 g/mol
Melting Pt: 186 °C
MDL Number: MFCD00016867
CAS Number: 29654-55-5
EINECS: 249-751-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a90ac1e467b26fba4210647ca1bdf896dbc88b29&vid=a90ac1e467b26fba4210647ca1bdf896dbc88b29&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR