3,5-Di-tert-butyl-2-hydroxybenzaldehyde ≥98.0%

Supplier: TCI America

Synonyms: 3,5-Di-tert-butylsalicylaldehyde

D2646-25G D2646-5G
TCD2646-25GEA 67.78 CAD
TCD2646-25G TCD2646-005G
3,5-Di-tert-butyl-2-hydroxybenzaldehyde ≥98.0%
3,5-Di-tert-butyl-2-hydroxybenzaldehyde

CAS Number: 37942-07-7
MDL Number: MFCD00191998
Molecular Formula: C15H22O2
Molecular Weight: 234.34
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 63


Other Product Names / Synonyms: 3,5-Di-tert-butyl-2-hydroxybenzaldehyde

Formula: C₁₅H₂₂O₂
MW: 234.34 g/mol
Melting Pt: 63 °C
MDL Number: MFCD00191998
CAS Number: 37942-07-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=074ad8ddef15396b23dc348b6ea269e2b2224001&vid=074ad8ddef15396b23dc348b6ea269e2b2224001&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR