1,3-Diacetylbenzene ≥98.0%
Supplier: TCI America
Synonyms:
Benzene-1,3-bis(acetyl)
D2617-5G
D2617-1G
TCD2617-005GEA
176.85
CAD
TCD2617-005G
76219-006
1,3-Diacetylbenzene ≥98.0%
1,3-Diacetylbenzene
CAS Number: 6781-42-6
MDL Number: MFCD00008740
Molecular Formula: C10H10O2
Molecular Weight: 162.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Flash Point (°C): 110
Freezing point (°C): 29
Formula:
C₁₀H₁₀O₂ Boiling Pt: 280 °C (1013 hPa) Melting Pt: 72…79 °C Flash Pt: 110 °C |
MDL Number:
MFCD00008740 CAS Number: 6781-42-6 EINECS: 229-842-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...