2,5-Dichloroanisole

Supplier: TCI America

Synonyms: 1,4-Dichloro-2-methoxybenzene

D2605-5G D2605-25G
TCD2605-005GEA 145.36 CAD
TCD2605-005G TCD2605-025G
2,5-Dichloroanisole
2,5-Dichloroanisole

CAS Number: 1984-58-3
MDL Number: MFCD00061123
Molecular Formula: C7H6Cl2O
Molecular Weight: 177.02
Form: Crystal
Color: White
Flash Point (°C): 110


Other Product Names / Synonyms: 2,5-Dichloromethoxybenzene, 1,4-Dichloro-2-methoxybenzene

Formula: C₇H₆Cl₂O
Boiling Pt: 225…227 °C (1013 hPa)
Melting Pt: 24 °C
Density: 1.333 g/cm³ (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00061123
CAS Number: 1984-58-3
EINECS: 217-852-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01bdc537c3b662144c2e44a478b849225e3b452c&vid=517f7cc5ad113ccc2ce43a8fe89c1ccf252c86eb&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR