2',4'-Difluoropropiophenone ≥98.0%
Supplier: TCI America
D2598-25G
D2598-5G
TCD2598-25GEA
267.81
CAD
TCD2598-25G
TCD2598-005G
2',4'-Difluoropropiophenone ≥98.0%
2',4'-Difluoropropiophenone
CAS Number: 85068-30-0
MDL Number: MFCD00015507
Molecular Formula: C9H8F2O
Molecular Weight: 170.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 76
Specific Gravity (20/20): 1.20
Formula:
C₉H₈F₂O Boiling Pt: 52 °C (1013 hPa) Density: 1.20 (20 °C) Flash Pt: 76 °C |
MDL Number:
MFCD00015507 CAS Number: 85068-30-0 EINECS: 285-291-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...