3,5-Difluorobenzoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

D2596-25G D2596-5G
TCD2596-25GEA 211.72 CAD
TCD2596-25G TCD2596-005G
3,5-Difluorobenzoic acid ≥98.0% (by titrimetric analysis)
3,5-Difluorobenzoic acid

CAS Number: 455-40-3
MDL Number: MFCD00010323
Molecular Formula: C7H4F2O2
Molecular Weight: 158.10
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 122

Formula: C₇H₄F₂O₂
Boiling Pt: 242 °C (1013 hPa)
Melting Pt: 122 °C
Storage Temperature: Ambient
MDL Number: MFCD00010323
CAS Number: 455-40-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0918f64cbe685df59a23388f7ba43059a29105b&vid=cde4ce205df23a29c324b68f2e951206d63ee343&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR