3,5-Dimethoxybenzyl alcohol ≥98.0%

Supplier: TCI America

Synonyms: (3,5-Dimethoxyphenyl)methanol

D2594-25G D2594-5G
TCD2594-25GEA 162.74 CAD
TCD2594-25G TCD2594-005G
3,5-Dimethoxybenzyl alcohol ≥98.0%
3,5-Dimethoxybenzyl alcohol

CAS Number: 705-76-0
MDL Number: MFCD00004641
Molecular Formula: C9H12O3
Molecular Weight: 168.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 171
Melting point (°C): 50
Flash Point (°C): 110

Formula: C₉H₁₂O₃
MW: 168.19 g/mol
Boiling Pt: 171 °C
Melting Pt: 50 °C
Flash Pt: 110 °C
MDL Number: MFCD00004641
CAS Number: 705-76-0
EINECS: 211-888-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98795859116a5fbd8445ed82600108f3fc5724bd&vid=98795859116a5fbd8445ed82600108f3fc5724bd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR