4,6-Dihydroxy-2-methylpyrimidine ≥96.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-Methylpyrimidine-4,6-diol, 2-Methyl-4,6-pyrimidinediol

D2589-5G D2589-25G
TCD2589-005GEA 41.08 CAD
TCD2589-005G TCD2589-025G
4,6-Dihydroxy-2-methylpyrimidine ≥96.0% (by titrimetric analysis)
4,6-Dihydroxy-2-methylpyrimidine

CAS Number: 40497-30-1
MDL Number: MFCD00006104
Molecular Formula: C5H6N2O2
Molecular Weight: 126.12
Purity/Analysis Method: >96.0% (T)
Form: Crystal


Other Product Names / Synonyms: 2-Methyl-4,6-pyrimidinediol

Formula: C₅H₆N₂O₂
MW: 126.12 g/mol
MDL Number: MFCD00006104
CAS Number: 40497-30-1
EINECS: 254-941-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1f1753236372b9e562869e5e366f35a29e48e09b&vid=1ed1a9616d3318fb712ca0326c36e406745482da&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR