3,5-Diamino-1H-1,2,4-triazole ≥97.0%
Supplier: TCI America
Synonyms:
3,5-Diamino-1,2,4-triazole, Guanazole
D2587-25G
D2587-5G
TCD2587-025GEA
203.82
CAD
TCD2587-025G
TCD2587-005G
3,5-Diamino-1H-1,2,4-triazole ≥97.0%
3,5-Diamino-1H-1,2,4-triazole
CAS Number: 1455-77-2
MDL Number: MFCD00005233
Molecular Formula: C2H5N5
Molecular Weight: 99.10
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Other Product Names / Synonyms: Guanazole
Formula:
C₂H₅N₅ MW: 99.1 g/mol Boiling Pt: 425 Melting Pt: 205 |
MDL Number:
MFCD00005233 CAS Number: 1455-77-2 EINECS: 215-937-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...