3,5-Dibromopyridine ≥98.0%
Supplier: TCI America
|
Warning
|
D2573-25G
D2573-5G
TCD2573-025GEA
266.07
CAD
TCD2573-025G
TCD2573-005G
3,5-Dibromopyridine ≥98.0%
3,5-Dibromopyridine
CAS Number: 625-92-3
MDL Number: MFCD00014634
Molecular Formula: C5H3Br2N
Molecular Weight: 236.89
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 222
Melting point (°C): 111
Flash Point (°C): 110
Formula:
C₅H₃Br₂N Boiling Pt: 222 °C Melting Pt: 111 °C Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014634 CAS Number: 625-92-3 EINECS: 210-916-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...