4-(Dimethylamino)benzophenone ≥98.0%
Supplier: TCI America
D2561-25G
D2561-5G
TCD2561-025GEA
193.39
CAD
TCD2561-025G
TCD2561-5G
4-(Dimethylamino)benzophenone ≥98.0%
4-(Dimethylamino)benzophenone
CAS Number: 530-44-9
MDL Number: MFCD00008311
Molecular Formula: C15H15NO
Molecular Weight: 225.29
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 250
Melting point (°C): 90
Formula:
C₁₅H₁₅NO Boiling Pt: 250 °C Melting Pt: 90 °C |
MDL Number:
MFCD00008311 CAS Number: 530-44-9 EINECS: 208-478-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...