2,5-Dimethoxytoluene ≥98.0%
Supplier: TCI America
|
Warning
|
D2552-5G
D2552-25G
TCD2552-005GEA
112.18
CAD
TCD2552-005G
TCD2552-025G
2,5-Dimethoxytoluene ≥98.0%
2,5-Dimethoxytoluene
CAS Number: 24599-58-4
MDL Number: MFCD00041901
Molecular Formula: C9H12O2
Molecular Weight: 152.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 218
Flash Point (°C): 102
Specific Gravity (20/20): 1.05
Formula:
C₉H₁₂O₂ Boiling Pt: 218 °C Melting Pt: 15 °C Density: 1.05 (20 °C) Flash Pt: 102 °C |
MDL Number:
MFCD00041901 CAS Number: 24599-58-4 EINECS: 246-343-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...