1,4-Dichloro-2-fluorobenzene ≥98.0%

Supplier: TCI America
Warning

D2513-25G D2513-5G
TCD2513-25GEA 274.92 CAD
TCD2513-25G TCD2513-005G
1,4-Dichloro-2-fluorobenzene ≥98.0%
1,4-Dichloro-2-fluorobenzene

CAS Number: 348-59-4
MDL Number: MFCD00060656
Molecular Formula: C6H3Cl2F
Molecular Weight: 164.99
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 168
Flash Point (°C): 65
Specific Gravity (20/20): 1.40

Formula: C₆H₃Cl₂F
Boiling Pt: 168 °C
Melting Pt: 4 °C
Density: 1.40 (20 °C)
Flash Pt: 65 °C
MDL Number: MFCD00060656
CAS Number: 348-59-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3a658bfa54706f55d9d56f6fa5ec7ea48f1107c&vid=08d6f3b85ec3a3982698fc57a28fae14636f4c9c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR