2,6-Difluorobenzyl alcohol ≥97.0%

Supplier: TCI America
Warning

Synonyms: (2,6-Difluorophenyl)methanol

D2507-5G D2507-25G
TCD2507-005GEA 72.68 CAD
TCD2507-005G TCD2507-025G
2,6-Difluorobenzyl alcohol ≥97.0%
2,6-Difluorobenzyl alcohol

CAS Number: 19064-18-7
MDL Number: MFCD00004603
Molecular Formula: C7H6F2O
Molecular Weight: 144.12
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 88
Flash Point (°C): 88
Specific Gravity (20/20): 1.30

Formula: C₇H₆F₂O
MW: 144.12 g/mol
Boiling Pt: 88 °C
Density: 1.30 (20 °C)
Flash Pt: 88 °C
MDL Number: MFCD00004603
CAS Number: 19064-18-7
EINECS: 242-792-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=482a3f6ed263a17ea794e3cb65e57ef9174e03a6&vid=482a3f6ed263a17ea794e3cb65e57ef9174e03a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR