2',5'-Dichloroacetophenone ≥98.0%

Supplier: TCI America

D2502-25G D2502-5G
TCD2502-025GEA 51.19 CAD
TCD2502-025G TCD2502-005G
2',5'-Dichloroacetophenone ≥98.0%
2',5'-Dichloroacetophenone

CAS Number: 2476-37-1
MDL Number: MFCD00000607
Molecular Formula: C8H6Cl2O
Molecular Weight: 189.04
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 128
Melting point (°C): 14
Flash Point (°C): 110
Specific Gravity (20/20): 1.33

Formula: C₈H₆Cl₂O
MW: 189.04 g/mol
Boiling Pt: 128 °C
Melting Pt: 14
Density: 1.33 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00000607
CAS Number: 2476-37-1
EINECS: 219-605-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=448325df5a05fe980d6d1edfb68e3d74caed01fd&vid=448325df5a05fe980d6d1edfb68e3d74caed01fd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR