2,4-Dichlorophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: Dihydroxy(2,4-dichlorophenyl)borane, 2,4-Dichlorobenzeneboronic acid, (2,4-Dichlorophenyl)metaboric acid

D2494-5G D2494-25G D2494-1G
TCD2494-5GEA 112.18 CAD
TCD2494-5G TCD2494-25G TCD2494-001G
2,4-Dichlorophenylboronic acid (contains varying amounts of Anhydride)
2,4-Dichlorophenylboronic acid

CAS Number: 68716-47-2
MDL Number: MFCD00013930
Molecular Formula: C6H5BCl2O2
Molecular Weight: 190.81
Form: Crystal
Color: White
Melting point (°C): 249


Other Product Names / Synonyms: 2,4-Dichlorobenzeneboronic Acid

Formula: C₆H₅BCl₂O₂
MW: 190.82 g/mol
Melting Pt: 249 °C
MDL Number: MFCD00013930
CAS Number: 68716-47-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=579a614be5706c4b5e37efe23b8ee4aca14fba47&vid=579a614be5706c4b5e37efe23b8ee4aca14fba47&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR