4,4'-Dipyridyl disulfide ≥97.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
4,4-Dipyridyl Disulfide, 4,4-Dipyridyl Disulphide, 4,4'-Dithiodipyridine
CAS Number: 2645-22-9
MDL Number: MFCD00006423
Molecular Formula: C10H8N2S2
Molecular Weight: 220.31
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 75
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Disulfides, Condensation & Active Esterification, Bioscience, Functional Group Blocking Reagents
Other Product Names / Synonyms: 4,4'-Dithiodipyridine
Formula:
C₁₀H₈N₂S₂ MW: 220.32 g/mol Boiling Pt: 348 °C (1013 hPa) Melting Pt: 75 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006423 CAS Number: 2645-22-9 EINECS: 220-158-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...