2,6-Dichloropurine ≥97.0%

Supplier: TCI America

Synonyms: 2,6-Dichloro-9H-purine

D2470-25G D2470-1G D2470-5G
TCD2470-25GEA 287.56 CAD
TCD2470-25G TCD2470-001G TCD2470-005G
2,6-Dichloropurine ≥97.0%
2,6-Dichloropurine

CAS Number: 5451-40-1
MDL Number: MFCD00077725
Molecular Formula: C5H2Cl2N4
Molecular Weight: 189.00
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal

Formula: C₅H₂Cl₂N₄
Melting Pt: 181 °C
MDL Number: MFCD00077725
CAS Number: 5451-40-1
EINECS: 226-681-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f0ead64c12e9f0e317ca3b98b69c858b7d814080&vid=f0ead64c12e9f0e317ca3b98b69c858b7d814080&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR