2,6-Difluorobenzaldehyde ≥98.0%

Supplier: TCI America

D2452-5G D2452-25G
TCD2452-005GEA 77.42 CAD
TCD2452-005G TCD2452-025G
2,6-Difluorobenzaldehyde ≥98.0%
2,6-Difluorobenzaldehyde

CAS Number: 437-81-0
MDL Number: MFCD00010293
Molecular Formula: C7H4F2O
Molecular Weight: 142.10
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 85
Flash Point (°C): 73
Freezing point (°C): 20
Specific Gravity (20/20): 1.33

Formula: C₇H₄F₂O
MW: 142.1 g/mol
Boiling Pt: 85 °C
Melting Pt: 15…17 °C
Density: 1.33 (20 °C)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00010293
CAS Number: 437-81-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c22364385985eee25d2faba8165807c9ab2be63b&vid=c3ac881cf63b2841b7a92d0544e4decceeaf2937&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR