4,4'-Di-tert-butylbiphenyl ≥98.0%
Supplier: TCI America
Synonyms:
1-tert-butyl-4-(4-tert-butylphenyl)benzene, 4,4'-Di-tert-butyl-1,1'-biphenyl
D2450-25G
D2450-5G
TCD2450-025GEA
141.47
CAD
TCD2450-025G
TCD2450-005G
4,4'-Di-tert-butylbiphenyl ≥98.0%
4,4'-Di-tert-butylbiphenyl
CAS Number: 1625-91-8
MDL Number: MFCD00008834
Molecular Formula: C20H26
Molecular Weight: 266.43
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 192
Melting point (°C): 128
Formula:
C₂₀H₂₆ MW: 266.43 g/mol Boiling Pt: 192 °C Melting Pt: 128 °C Density: 0.9691 g/cm³ (20 °C) |
MDL Number:
MFCD00008834 CAS Number: 1625-91-8 EINECS: 216-615-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...