1,3-Di-tert-butylbenzene ≥98.0%

Supplier: TCI America

D2431-5ML D2431-1ML
TCD2431-005MLEA 402.73 CAD
TCD2431-005ML TCD2431-1ML
1,3-Di-tert-butylbenzene ≥98.0%
1,3-Di-tert-butylbenzene

CAS Number: 1014-60-4
MDL Number: MFCD00008830
Molecular Formula: C14H22
Molecular Weight: 190.33
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 107
Melting point (°C): 11
Flash Point (°C): 84
Specific Gravity (20/20): 0.86

Formula: C₁₄H₂₂
Boiling Pt: 107 °C
Melting Pt: 11 °C
Density: 0.86 (20 °C)
Flash Pt: 84 °C
MDL Number: MFCD00008830
CAS Number: 1014-60-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d21e3b5fe6c134166d218eda7ef1059ed6134892&vid=d21e3b5fe6c134166d218eda7ef1059ed6134892&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR