2,6-Di-tert-butyl-4-methylpyridine ≥98.0%

Supplier: TCI America
Warning

Synonyms: 2,6-Di-Tert-butyl-4-methylpyridine

D2419-25G D2419-5G
TCD2419-25GEA 692.04 CAD
TCD2419-25G TCD2419-005G
2,6-Di-tert-butyl-4-methylpyridine ≥98.0%
2,6-Di-tert-butyl-4-methylpyridine

CAS Number: 38222-83-2
MDL Number: MFCD00006305
Molecular Formula: C14H23N
Molecular Weight: 205.35
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 226
Melting point (°C): 34
Flash Point (°C): 84
Storage Temperature: 0-10°C


Other Product Names / Synonyms: DTBMP

Formula: C₁₄H₂₃N
MW: 205.34 g/mol
Boiling Pt: 226 °C
Melting Pt: 34 °C
Flash Pt: 84 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006305
CAS Number: 38222-83-2
EINECS: 253-834-4

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR