3,4-Difluorobenzyl alcohol

Supplier: TCI America
Warning

D2415-25G D2415-5G
TCD2415-25GEA 208.56 CAD
TCD2415-25G TCD2415-005G
3,4-Difluorobenzyl alcohol
3,4-Difluorobenzyl alcohol

CAS Number: 85118-05-4
MDL Number: MFCD00010628
Molecular Formula: C7H6F2O
Molecular Weight: 144.12
Form: Clear Liquid
Color: Pale Yellow
Flash Point (°C): 97
Specific Gravity (20/20): 1.29

Formula: C₇H₆F₂O
Boiling Pt: 197 °C (1013 hPa)
Melting Pt: 9 °C
Density: 1.29 (20 °C)
Flash Pt: 97 °C
MDL Number: MFCD00010628
CAS Number: 85118-05-4
EINECS: 285-657-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e0525e57b907b27157bc6cff291061157b96e354&vid=e0525e57b907b27157bc6cff291061157b96e354&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR