2,6-Dibromo-4-nitroaniline ≥98.0%

Supplier: TCI America
Warning

D2414-25G D2414-500G
TCD2414-025GEA 79.32 CAD
TCD2414-025G TCD2414-500G
2,6-Dibromo-4-nitroaniline ≥98.0%
2,6-Dibromo-4-nitroaniline

CAS Number: 827-94-1
MDL Number: MFCD00007639
Molecular Formula: C6H4Br2N2O2
Molecular Weight: 295.92
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 207

Formula: C₆H₄Br₂N₂O₂
MW: 295.92 g/mol
Melting Pt: 207 °C
Storage Temperature: Ambient
MDL Number: MFCD00007639
CAS Number: 827-94-1
EINECS: 212-577-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=573d9a3c486bcafa8cb66d01ff38c599d4f9d61a&vid=8e363223bc97a7e3eac493b1c91d858ced3f2fa9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR