Diphenylsilane ≥97.0%

Supplier: TCI America
Warning

D2406-5G D2406-25G
TCD2406-5GEA 99.51 CAD
TCD2406-5G TCD2406-025G
Diphenylsilane ≥97.0%
Diphenylsilane

CAS Number: 775-12-2
MDL Number: MFCD00003002
Molecular Formula: C12H12Si
Molecular Weight: 184.31
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 127
Flash Point (°C): 98
Specific Gravity (20/20): 1.00

Formula: C₁₂H₁₂Si
MW: 184.31 g/mol
Boiling Pt: 127 °C
Density: 1.00 (20 °C)
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00003002
CAS Number: 775-12-2
EINECS: 212-271-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea1204ae0fc5fd1739c10204a69fd067d024c3ee&vid=ea1204ae0fc5fd1739c10204a69fd067d024c3ee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR