2,4-Dimethylbenzaldehyde ≥97.0%

Supplier: TCI America

D2392-25G D2392-500G
TCD2392-025GEA 77.42 CAD
TCD2392-025G TCD2392-500G
2,4-Dimethylbenzaldehyde ≥97.0%
2,4-Dimethylbenzaldehyde

CAS Number: 15764-16-6
MDL Number: MFCD00003340
Molecular Formula: C9H10O
Molecular Weight: 134.18
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 98
Flash Point (°C): 89
Specific Gravity (20/20): 1.02


Other Product Names / Synonyms: m-Xylene-4-carboxaldehyde

Formula: C₉H₁₀O
MW: 134.18 g/mol
Boiling Pt: 98 °C
Melting Pt: –9 °C
Density: 1.02 (20 °C)
Flash Pt: 89 °C
Storage Temperature: Ambient
MDL Number: MFCD00003340
CAS Number: 15764-16-6
EINECS: 239-856-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97d0c7188bd2755ba5b12808caec76988d58198b&vid=60463b13a8d73d69f721b48b93f8141538338255&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR