3,5-Di-tert-butyl-4-hydroxybenzoic acid ≥98.0%

Supplier: TCI America

D2381-250G D2381-25G
TCD2381-250GEA 419.81 CAD
TCD2381-250G TCD2381-025G
3,5-Di-tert-butyl-4-hydroxybenzoic acid ≥98.0%
3,5-Di-tert-butyl-4-hydroxybenzoic acid

CAS Number: 1421-49-4
MDL Number: MFCD00008827
Molecular Formula: C15H22O3
Molecular Weight: 250.34
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 210

Formula: C₁₅H₂₂O₃
MW: 250.34 g/mol
Boiling Pt: 374
Melting Pt: 210 °C
MDL Number: MFCD00008827
CAS Number: 1421-49-4
EINECS: 215-823-2

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR