2,5-Dibromo-3,4-dinitrothiophene ≥98.0%

Supplier: TCI America

D2371-5G D2371-1G
TCD2371-5GEA 203.82 CAD
TCD2371-5G TCD2371-001G
2,5-Dibromo-3,4-dinitrothiophene ≥98.0%
2,5-Dibromo-3,4-dinitrothiophene

CAS Number: 52431-30-8
MDL Number: MFCD00015537
Molecular Formula: C4Br2N2O4S
Molecular Weight: 331.92
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 137

Formula: C₄Br₂N₂O₄S
Melting Pt: 137 °C
MDL Number: MFCD00015537
CAS Number: 52431-30-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e0d433154f6ba76c26316dcc43f48cea1b462fc4&vid=e0d433154f6ba76c26316dcc43f48cea1b462fc4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR