1,4-Dibromonaphthalene ≥98.0%

Supplier: TCI America
Warning

D2359-25G D2359-5G
TCD2359-25GEA 335.59 CAD
TCD2359-25G TCD2359-005G
1,4-Dibromonaphthalene ≥98.0%
1,4-Dibromonaphthalene

CAS Number: 83-53-4
MDL Number: MFCD00041823
Molecular Formula: C10H6Br2
Molecular Weight: 285.97
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 82
Flash Point (°C): 100

Formula: C₁₀H₆Br₂
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 82 °C
Flash Pt: 100 °C
MDL Number: MFCD00041823
CAS Number: 83-53-4
EINECS: 201-484-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=866aa13ef97a553ac3c903e48969307d76d63720&vid=866aa13ef97a553ac3c903e48969307d76d63720&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR