3,5-Dibromobenzoic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Warning

D2353-25G D2353-5G
TCD2353-010GEA 150.34 CAD
TCD2353-010G TCD2353-25G TCD2353-5G
3,5-Dibromobenzoic acid ≥97.0% (by GC, titration analysis)
3,5-Dibromobenzoic acid

CAS Number: 618-58-6
MDL Number: MFCD00051758
Molecular Formula: C7H4Br2O2
Molecular Weight: 279.92
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 220

Formula: C₇H₄Br₂O₂
Melting Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00051758
CAS Number: 618-58-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f655a3c59db623edfd6224079b75b93e0a296a42&vid=f655a3c59db623edfd6224079b75b93e0a296a42&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR